missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethyl-2,4-dimethylpyrrole, TCI America™

Manufacturer:  TCI America E05551G

 View more versions of this product

Catalog No. E05551G



Specifications

Specifications

3-Ethyl-2,4-dimethylpyrrole
1g
C8H13N
2,4-dimethyl-3-ethylpyrrole, kryptopyrrole, kryptopyrrol, 3-ethyl-2,4-dimethylpyrrole, cryptopyrrole, 2,4-dimethyl-3-ethyl-1h-pyrrole, 1h-pyrrole, 3-ethyl-2,4-dimethyl, pyrrole, 3-ethyl-2,4-dimethyl, acmc-209kvh, 5-20-05-00099 beilstein handbook reference
CCC1=C(NC=C1C)C
123.199
123.20
≥97.0% (GC)
Yellow
517-22-6
MFCD00005222
ZEBBLOXDLGIMEG-UHFFFAOYSA-N
3-ethyl-2,4-dimethyl-1H-pyrrole
10600
Clear Liquid at 20°C

Chemical Identifiers

517-22-6
123.199
ZEBBLOXDLGIMEG-UHFFFAOYSA-N
10600
CCC1=C(NC=C1C)C
C8H13N
MFCD00005222
2,4-dimethyl-3-ethylpyrrole, kryptopyrrole, kryptopyrrol, 3-ethyl-2,4-dimethylpyrrole, cryptopyrrole, 2,4-dimethyl-3-ethyl-1h-pyrrole, 1h-pyrrole, 3-ethyl-2,4-dimethyl, pyrrole, 3-ethyl-2,4-dimethyl, acmc-209kvh, 5-20-05-00099 beilstein handbook reference
3-ethyl-2,4-dimethyl-1H-pyrrole
Safety and Handling

Safety and Handling

RTECSNumber : UX9629500

TSCA : No

Recommended Storage : Refrigerator

Documents