missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethyl-1-pentyn-3-ol, TCI America™

Manufacturer:  TCI America E02735ML

 View more versions of this product

Catalog No. E02735ML



Specifications

Specifications

3-Ethyl-1-pentyn-3-ol
Yellow
6285-06-9
MFCD00014392
3-ethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3-ethyl, diethylethynylcarbinol, 3-pentanol, 3-ethynyl, diaethyl-aethenyl-carbinol german, 3-aethyl-pentin-1-ol-3, 3-aethyl-pentin-1-ol-3 german, d iethylethynylcarbinol, acmc-1bagf, diaethyl-aethenyl-carbinol
CCC(CC)(C#C)O
112.17
112.17
≥97.0% (GC)
138°C
5mL
C7H12O
1987
PUNRPAWKFTXZIW-UHFFFAOYSA-N
3-ethylpent-1-yn-3-ol
22698
Clear Liquid at 20°C

Chemical Identifiers

6285-06-9
112.17
PUNRPAWKFTXZIW-UHFFFAOYSA-N
22698
CCC(CC)(C#C)O
C7H12O
MFCD00014392
3-ethyl-1-pentyn-3-ol, 1-pentyn-3-ol, 3-ethyl, diethylethynylcarbinol, 3-pentanol, 3-ethynyl, diaethyl-aethenyl-carbinol german, 3-aethyl-pentin-1-ol-3, 3-aethyl-pentin-1-ol-3 german, d iethylethynylcarbinol, acmc-1bagf, diaethyl-aethenyl-carbinol
3-ethylpent-1-yn-3-ol
Safety and Handling

Safety and Handling

RTECSNumber : SC4857000

TSCA : No

Documents