missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-1-pentyn-3-ol, 98%

$135.92 - $135.92

Chemical Identifiers

CAS 6285-06-9
Molecular Formula C7H12O
Formula Weight 112.17
MDL Number MFCD00014392
UN Number UN1987
Beilstein 1740944
Synonym Diethyl ethynyl carbinol
Chemical Name or Material 3-Ethyl-1-pentyn-3-ol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0633506 SDS Alfa Aesar™
L0633506
5g Each for $135.92
AAL0633514 SDS Alfa Aesar™
L0633514
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAL0633522 SDS Alfa Aesar™
L0633522
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
Description & Specifications

Chemical Identifiers

6285-06-9
112.17
UN1987
Diethyl ethynyl carbinol
C7H12O
MFCD00014392
1740944
3-Ethyl-1-pentyn-3-ol

Specifications

38°C (100°F)
5g
C7H12O
MFCD00014392
1740944
3-Ethyl-1-pentyn-3-ol
138° to 140°C
6285-06-9
112.17
UN1987
Diethyl ethynyl carbinol
98%

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

RTECSNumber : SC4857000