Learn More
3-Ethyl-1-pentyn-3-ol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0633506
Description
3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
Specifications
3-Ethyl-1-pentyn-3-ol | |
0.873 | |
38°C (100°F) | |
1.438 | |
5 g | |
1740944 | |
Not miscible in water. | |
CC(CCO)O | |
90.12 | |
CHEBI:52687 | |
98% |
6285-06-9 | |
138°C to 140°C | |
C4H10O2 | |
MFCD00014392 | |
UN1987 | |
r---1,3-butanediol, r-butane-1,3-diol, 3r-butane-1,3-diol, r-1,3-butanediol, 1,3-butanediol, 3r, r---1,3-butylene glycol, r---butane-1,3-diol, 1,3-butanediol, r | |
PUPZLCDOIYMWBV-UHFFFAOYNA-N | |
(3R)-butane-1,3-diol | |
637497 | |
112.17 |
Chemical Identifiers
6285-06-9 | |
MFCD00014392 | |
CHEBI:52687 |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 228-512-1
RTECSNumber : SC4857000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only