Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethyl-1-pentyn-3-ol, 98%


Quantity 100g

Manufacturer: alfa aesar™  L0633522

 View more versions of this product

Catalog No. AAL0633522

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 112.17
Synonym Diethyl ethynyl carbinol
Quantity 100g
Chemical Name or Material 3-Ethyl-1-pentyn-3-ol
Assay 98%
MDL Number MFCD00014392
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Molecular Formula C7H12O
CAS 6285-06-9
Beilstein 1740944
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SC4857000
Boiling Point 138° to 140°C
UN Number UN1987
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Flash Point 38°C (100°F)

Specifications

Chemical Name or Material 3-Ethyl-1-pentyn-3-ol
CAS 6285-06-9
Molecular Formula C7H12O
Formula Weight 112.17
MDL Number MFCD00014392
UN Number UN1987
Beilstein 1740944
Synonym Diethyl ethynyl carbinol

Applications
3-Ethyl-1-pentyn-3-ol is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Accessories

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.

SDS & Safety Documents