Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxysalicylaldehyde, 97%

$46.32 - $181.64

Chemical Identifiers

Chemical Name or Material 3-Ethoxysalicylaldehyde
CAS 492-88-6
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00003323
Synonym 3-Ethoxy-2-hydroxybenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2070218

  SDS
alfa aesar™
B2070218
50g Each for $46.32

AAB2070230

  SDS
alfa aesar™
B2070230
250g Each for $181.64
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material 3-Ethoxysalicylaldehyde
TSCA TSCA
Assay 97%
Molecular Formula C9H10O3
MDL Number MFCD00003323
CAS 492-88-6
Synonym 3-Ethoxy-2-hydroxybenzaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CU6123000
Boiling Point 263° to 264°C
Melting Point 66° to 68°C
Quantity 50g

Applications
3-Ethoxysalicylaldehyde is employed in the preparation of Schiff base ligand through condensation with trans-1,2-diaminocyclohexane. It is also useful for the preparation of substituted salen-type Schiff base ligands. Further, it is used to prepare 2-ethoxy-6-[(3-methylaminopropylimino)methyl]phenol by reaction with N-methylpropane-1,3-diamine.

Solubility
Soluble in cold water. Insoluble in methanol, ethanol, acetone and acetonitrile

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.