Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxypropylamine, 99%

Chemical Identifiers

Chemical Name or Material 3-Ethoxypropylamine
CAS 6291-85-6
Molecular Formula C5H13NO
Linear Formula CH3CH2O(CH2)3NH2
Formula Weight 103.16
MDL Number MFCD00008221
UN Number UN2733
Beilstein 1732287
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2369422

 
alfa aesar™
B2369422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2369436

 
alfa aesar™
B2369436
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 103.16
Chemical Name or Material 3-Ethoxypropylamine
TSCA TSCA
Assay 99%
Molecular Formula C5H13NO
Linear Formula CH3CH2O(CH2)3NH2
MDL Number MFCD00008221
CAS 6291-85-6
UN Number UN2733
Beilstein 1732287
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
RTECS Number UI2700000
Flash Point 32°C (91°F)
Boiling Point 136° to 138°C
Quantity 100g

Applications
It is employed as an intermediate used in the production of dyestuff and cosmetics. It is also used to partially open the rigid oxetane chain and to identify Hsp70 modulators.

Solubility
Miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.