Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxypropionitrile, 95%

$74.97 - $342.89

Chemical Identifiers

Chemical Name or Material 3-Ethoxypropionitrile
CAS 2141-62-0
Molecular Formula C5H9NO
Linear Formula CH3CH2OCH2CH2CN
Formula Weight 99.14g/mol
MDL Number MFCD00001959
Beilstein 741924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA142430F

 
alfa aesar™
A14243-0F
2500mL Each for $342.89

AAA14243AP

 
alfa aesar™
A14243-AP
500mL Each for $74.97
Description & Specifications

Specifications

Boiling Point 171° to 173°C
Quantity 2500mL
Flash Point 63°C (145°F)
Chemical Name or Material 3-Ethoxypropionitrile
CAS 2141-62-0
Assay 95%
Molecular Formula C5H9NO
Linear Formula CH3CH2OCH2CH2CN
Formula Weight 99.14g/mol
MDL Number MFCD00001959
Beilstein 741924
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TZ4680000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.