missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxypiperidine, 96%

$107.30 - $369.43

Chemical Identifiers

CAS 88536-17-8
Molecular Formula C7H15NO
Formula Weight 129.2
MDL Number MFCD05022464
Chemical Name or Material 3-Ethoxypiperidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5095703 SDS Alfa Aesar™
H5095703
1g Each for $369.43
AAH50957MD SDS Alfa Aesar™
H50957MD
250mg Each for $107.30
Specifications Description & Specifications

Chemical Identifiers

88536-17-8
129.2
3-Ethoxypiperidine
C7H15NO
MFCD05022464

Specifications

1g
C7H15NO
MFCD05022464
96%
88536-17-8
129.2
3-Ethoxypiperidine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning