Learn More
3-Ethoxymethacrolein, 95%, Tech., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 213990100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Ethoxymethacrolein | |
42588-57-8 | |
100.0 | |
0.9600g/mL | |
35°C | |
94% min. (GC) | |
C6H10O2 | |
C2H5OCH=C(CH3)CHO | |
10 g | |
0.96 | |
KDOAHVPFGIYCEU-AATRIKPKSA-N | |
(Z)-3-ethoxy-2-methylprop-2-enal | |
12278948 | |
95% | |
Liquid |
tech., 95% | |
94.0 | |
Yellow | |
78.0°C to 81.0°C (14.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4792 to 1.4812 | |
MFCD00006975 | |
01, IV, 4082 | |
3-ethoxymethacrolein, 3-ethoxy-2-methylacrylaldehyde, 3-ethoxy-2-methyl-2-propenal, 2e-3-ethoxy-2-methylprop-2-enal, 3-ethoxy-2-methylpropenal, 3-ethoxy-2-methylacroleine, e-2-methyl-3-ethoxypropenal, 2-propenal, 3-ethoxy-2-methyl, e-3-ethoxy-2-methylprop-2-enal | |
CCO\C=C(/C)C=O | |
114.14 | |
114.14 | |
Technical |
Chemical Identifiers
42588-57-8 | |
114.14 | |
KDOAHVPFGIYCEU-AATRIKPKSA-N | |
12278948 | |
CCO\C=C(/C)C=O |
C6H10O2 | |
MFCD00006975 | |
3-ethoxymethacrolein, 3-ethoxy-2-methylacrylaldehyde, 3-ethoxy-2-methyl-2-propenal, 2e-3-ethoxy-2-methylprop-2-enal, 3-ethoxy-2-methylpropenal, 3-ethoxy-2-methylacroleine, e-2-methyl-3-ethoxypropenal, 2-propenal, 3-ethoxy-2-methyl, e-3-ethoxy-2-methylprop-2-enal | |
(Z)-3-ethoxy-2-methylprop-2-enal |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Warning
EINECSNumber : 255-899-4
TSCA : TSCA
RUO – Research Use Only