Please login to your online account to display your discounted pricing

(3-Ethoxycarbonylpropyl)triphenylphosphonium bromide, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (3-Ethoxycarbonylpropyl)triphenylphosphonium bromide
CAS 50479-11-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C24H26BrO2P
Formula Weight 457.35
MDL Number MFCD00274610
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389770250

  SDS
thermo fisher scientific
AC389770250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 457.35
Physical Form Powder
Quantity 25g
Color White
Melting Point 167° to 170°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material (3-Ethoxycarbonylpropyl)triphenylphosphonium bromide
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C24H26BrO2P
MDL Number MFCD00274610
CAS 50479-11-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Contact with water liberates toxic gas.

For C-C Bond Formation (Olefination)

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Contact with water liberates toxic gas.