Please login to your online account to display your discounted pricing

3-Ethoxycarbonylphenylboronic acid, 97%, ACROS Organics™

$60.19 - $60.19

Chemical Identifiers

Chemical Name or Material 3-Ethoxycarbonylphenylboronic acid
CAS 4334-87-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H11BO4
Formula Weight 193.99
MDL Number MFCD02179444
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC359305000

  SDS
thermo fisher scientific
35930-5000
Glass bottle 500mg Each for $60.19
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 193.99
Quantity 500mg
Color White
TSCA TSCA
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C9H11BO4
CAS 4334-87-6
Chemical Name or Material 3-Ethoxycarbonylphenylboronic acid
MDL Number MFCD02179444
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.