missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Ethoxycarbonyl)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

CAS 269410-00-6
Molecular Formula C15H21BO4
Molecular Weight (g/mol) 276.14
InChI Key RQKGHIGGSFNVGW-UHFFFAOYSA-N
Synonym 3-Carboethoxyphenylboronic acid pinacol ester
IUPAC Name ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES CCOC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445020010
SDS View Documents
Thermo Fisher Scientific
AC445020010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445020050
SDS View Documents
Thermo Fisher Scientific
AC445020050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

269410-00-6
276.14
3-Carboethoxyphenylboronic acid pinacol ester
CCOC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
C15H21BO4
RQKGHIGGSFNVGW-UHFFFAOYSA-N
ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Specifications

110°C
41°C to 43°C
269410-00-6
C15H21BO4
RQKGHIGGSFNVGW-UHFFFAOYSA-N
ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
276.14
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Glass Bottle
1g
96% min. (GC)
3-Carboethoxyphenylboronic acid pinacol ester
CCOC(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
276.14
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning

SDS