missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Ethoxycarbonyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0949503

 View more versions of this product

Catalog No. AAL0949503



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(Ethoxycarbonyl)phenyl isocyanate
1.154
>110°C (230°F)
1.531
1 g
7636626
3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester
CCOC(=O)C1=CC(=CC=C1)N=C=O
191.19
191.19
67531-68-4
213°C
C10H9NO3
MFCD00013865
UN2206
Moisture sensitive
UPXGGIRFDZENRD-UHFFFAOYSA-N
ethyl 3-isocyanatobenzoate
589806
97%

Chemical Identifiers

67531-68-4
191.19
UPXGGIRFDZENRD-UHFFFAOYSA-N
589806
CCOC(=O)C1=CC(=CC=C1)N=C=O
C10H9NO3
MFCD00013865
3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester
ethyl 3-isocyanatobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H331-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only