missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Ethoxycarbonyl-2-piperidone 98.0+%, TCI America™
$92.90 - $273.73
Chemical Identifiers
CAS | 3731-16-6 |
---|---|
Molecular Formula | C8H13NO3 |
Molecular Weight (g/mol) | 171.196 |
MDL Number | MFCD00006038 |
InChI Key | DUMNOWYWTAYLJN-UHFFFAOYSA-N |
Synonym | Ethyl 2-Piperidone-3-carboxylate, Ethyl 2-Oxo-3-piperidinecarboxylate, 2-Oxo-3-piperidinecarboxylic Acid Ethyl Ester |
PubChem CID | 97773 |
IUPAC Name | ethyl 2-oxopiperidine-3-carboxylate |
SMILES | CCOC(=O)C1CCCNC1=O |
Chemical Identifiers
3731-16-6 | |
171.196 | |
DUMNOWYWTAYLJN-UHFFFAOYSA-N | |
97773 | |
CCOC(=O)C1CCCNC1=O |
C8H13NO3 | |
MFCD00006038 | |
Ethyl 2-Piperidone-3-carboxylate, Ethyl 2-Oxo-3-piperidinecarboxylate, 2-Oxo-3-piperidinecarboxylic Acid Ethyl Ester | |
ethyl 2-oxopiperidine-3-carboxylate |
Specifications
3731-16-6 | |
White-Yellow | |
MFCD00006038 | |
Ethyl 2-Piperidone-3-carboxylate, Ethyl 2-Oxo-3-piperidinecarboxylate, 2-Oxo-3-piperidinecarboxylic Acid Ethyl Ester | |
CCOC(=O)C1CCCNC1=O | |
171.196 | |
171.20 | |
Crystalline Powder |
82°C | |
C8H13NO3 | |
5 g | |
DUMNOWYWTAYLJN-UHFFFAOYSA-N | |
ethyl 2-oxopiperidine-3-carboxylate | |
97773 | |
≥98.0% (HPLC) | |
3-Ethoxycarbonyl-2-piperidone |
Safety and Handling
TSCA : No