missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxybenzhydrazide, 98%

$74.65 - $258.59

Chemical Identifiers

CAS 27830-16-6
Molecular Formula C9H12N2O2
Formula Weight 180.21
MDL Number MFCD00017063
Beilstein 2577763
Chemical Name or Material 3-Ethoxybenzhydrazide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0633003 SDS Alfa Aesar™
L0633003
1g Each for $74.65
AAL0633006 SDS Alfa Aesar™
L0633006
5g Each for $258.59
Specifications

Description

Applications
3-Ethoxybenzhydrazide is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

27830-16-6
180.21
2577763
C9H12N2O2
MFCD00017063
3-Ethoxybenzhydrazide

Specifications

27830-16-6
180.21
2577763
1g
98%
C9H12N2O2
MFCD00017063
101° to 104°C
3-Ethoxybenzhydrazide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning