Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxybenzhydrazide, 98%


Quantity 1g

Manufacturer: alfa aesar™  L0633003

 View more versions of this product

Catalog No. AAL0633003

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 180.21
Chemical Name or Material 3-Ethoxybenzhydrazide
MDL Number MFCD00017063
Assay 98%
Molecular Formula C9H12N2O2
CAS 27830-16-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Beilstein 2577763
Melting Point 101° to 104°C

Specifications

Chemical Name or Material 3-Ethoxybenzhydrazide
CAS 27830-16-6
Molecular Formula C9H12N2O2
Formula Weight 180.21
MDL Number MFCD00017063
Beilstein 2577763

Applications
3-Ethoxybenzhydrazide is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents