missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethoxybenzenethiol, TCI America™

Manufacturer:  TCI America E07015G

 View more versions of this product

Catalog No. E07015G



Specifications

Specifications

3-Ethoxybenzenethiol
Yellow
86704-82-7
MFCD03094660
RTJMJGJLVSKSEB-UHFFFAOYSA-N
3-ethoxybenzenethiol
2736350
Clear Liquid at 20°C
239°C
5g
C8H10OS
3-Ethoxythiophenol
CCOC1=CC(=CC=C1)S
154.227
154.23
≥98.0% (GC)

Chemical Identifiers

86704-82-7
154.227
RTJMJGJLVSKSEB-UHFFFAOYSA-N
2736350
CCOC1=CC(=CC=C1)S
C8H10OS
MFCD03094660
3-Ethoxythiophenol
3-ethoxybenzenethiol
Safety and Handling

Safety and Handling

TSCA : No

Documents