Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxybenzeneboronic acid, 98%

$64.93 - $176.30

Chemical Identifiers

Chemical Name or Material 3-Ethoxybenzeneboronic acid
CAS 90555-66-1
Molecular Formula C8H11BO3
Formula Weight 165.99
MDL Number MFCD00274219
Synonym 3-Ethoxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2448503

  SDS
alfa aesar™
B2448503
1g Each for $64.93

AAB2448506

  SDS
alfa aesar™
B2448506
5g Each for $176.30
Description & Specifications

Specifications

Formula Weight 165.99
Chemical Name or Material 3-Ethoxybenzeneboronic acid
Assay 98%
Molecular Formula C8H11BO3
MDL Number MFCD00274219
CAS 90555-66-1
Synonym 3-Ethoxyphenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 142° to 146°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.