Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxybenzamide, 98%

$27.43 - $27.43

Chemical Identifiers

Chemical Name or Material 3-Ethoxybenzamide
CAS 55836-69-6
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD00007988
Beilstein 2638808
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0561603

  SDS
alfa aesar™
L0561603
1g Each for $27.43
Description & Specifications

Specifications

Formula Weight 165.19
Chemical Name or Material 3-Ethoxybenzamide
CAS 55836-69-6
Molecular Formula C9H11NO2
MDL Number MFCD00007988
Beilstein 2638808
Quantity 1g
Melting Point 137° to 140°C
Assay 98%