missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Ethoxybenzaldehyde 98.0+%, TCI America™
Supplier: TCI America E113325G
Specifications
3-Ethoxybenzaldehyde | |
Yellow | |
C9H10O2 | |
25 g | |
QZMGMXBYJZVAJN-UHFFFAOYSA-N | |
3-ethoxybenzaldehyde | |
89908 | |
≥98.0% (GC) |
22924-15-8 | |
245°C | |
MFCD00016606 | |
m-ethoxybenzaldehyde, benzaldehyde, 3-ethoxy, 3-ethoxy-benzaldehyde, benzaldehyde, m-ethoxy, 3-ethoxy benzaldehyde, 3-ethoxybenzaldehdye, pubchem13132, acmc-209fzo, intermediates-zcf02615, 3-ethoxybenzaldehyde | |
CCOC1=CC=CC(=C1)C=O | |
150.177 | |
150.18 | |
Liquid |
Chemical Identifiers
22924-15-8 | |
150.177 | |
QZMGMXBYJZVAJN-UHFFFAOYSA-N | |
89908 | |
CCOC1=CC=CC(=C1)C=O |
C9H10O2 | |
MFCD00016606 | |
m-ethoxybenzaldehyde, benzaldehyde, 3-ethoxy, 3-ethoxy-benzaldehyde, benzaldehyde, m-ethoxy, 3-ethoxy benzaldehyde, 3-ethoxybenzaldehdye, pubchem13132, acmc-209fzo, intermediates-zcf02615, 3-ethoxybenzaldehyde | |
3-ethoxybenzaldehyde |
Safety and Handling
EINECSNumber : (3)-2662
TSCA : No