missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethoxyacrylonitrile (cis- and trans- mixture), TCI America™

Manufacturer:  TCI America E0388250ML

 View more versions of this product

Catalog No. E0388250ML



Specifications

Specifications

3-Ethoxyacrylonitrile (cis- and trans- mixture)
Yellow
61310-53-0
MFCD00010193
HUPVIAINOSTNBJ-HWKANZROSA-N
(E)-3-ethoxyprop-2-enenitrile
5324714
Clear Liquid at 20°C
71°C
250mL
C5H7NO
2z-3-ethoxyprop-2-enenitrile, z-3-ethoxyprop-2-enenitrile, 2-propenenitrile, 3-ethoxy, z-3-ethoxy-2-propenenitrile
CCOC=CC#N
97.117
97.12
≥98.0% (GC)

Chemical Identifiers

61310-53-0
97.117
HUPVIAINOSTNBJ-HWKANZROSA-N
5324714
CCOC=CC#N
C5H7NO
MFCD00010193
2z-3-ethoxyprop-2-enenitrile, z-3-ethoxyprop-2-enenitrile, 2-propenenitrile, 3-ethoxy, z-3-ethoxy-2-propenenitrile
(E)-3-ethoxyprop-2-enenitrile
Safety and Handling

Safety and Handling

RTECSNumber : AT6475000

TSCA : No

Documents