Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Ethoxyacetanilide, 99+%

$126.22 - $126.22

Chemical Identifiers

Chemical Name or Material 3'-Ethoxyacetanilide
CAS 591-33-3
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00061110
Synonym 3-Acetophenetidide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2533914

  SDS
alfa aesar™
B2533914
25g Each for $126.22
Description & Specifications

Specifications

Formula Weight 179.22
Chemical Name or Material 3'-Ethoxyacetanilide
TSCA TSCA
Assay ≥99%
Molecular Formula C10H13NO2
MDL Number MFCD00061110
CAS 591-33-3
Synonym 3-Acetophenetidide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 95° to 98°C
Quantity 25g
RTECS Number AM4300000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.