missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Ethoxy-5-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$127.75 - $379.90
Chemical Identifiers
CAS | 850589-53-6 |
---|---|
Molecular Formula | C8H10BFO3 |
Molecular Weight (g/mol) | 183.97 |
MDL Number | MFCD07363779 |
InChI Key | OVVBLFYHZAMJIK-UHFFFAOYSA-N |
Synonym | 3-ethoxy-5-fluorobenzeneboronic acid, 3-ethoxy-5-fluorophenyl boronic acid, 5-ethoxy-3-fluorophenylboronic acid, boronic acid,b-3-ethoxy-5-fluorophenyl, 3-ethoxy-5-fluorophenylboronicacid, acmc-209q3b, 3-ethoxy-5-fluoro-benzeneboronic acid, 3-ethoxy-5-fluoro-phenyl boronic acid, 3-ethoxy-5-fluorobenzeneboronicacid, 3-ethoxy-5-fluoranyl-phenyl boronic acid |
PubChem CID | 44558175 |
IUPAC Name | (3-ethoxy-5-fluorophenyl)boronic acid |
SMILES | CCOC1=CC(F)=CC(=C1)B(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
E1205200MG
|
TCI America
E1205200MG |
200 mg |
Each for $127.75
|
|
|||||
E12051G
|
TCI America
E12051G |
1 g |
Each for $379.90
|
|
|||||
Chemical Identifiers
850589-53-6 | |
183.97 | |
OVVBLFYHZAMJIK-UHFFFAOYSA-N | |
44558175 | |
CCOC1=CC(F)=CC(=C1)B(O)O |
C8H10BFO3 | |
MFCD07363779 | |
3-ethoxy-5-fluorobenzeneboronic acid, 3-ethoxy-5-fluorophenyl boronic acid, 5-ethoxy-3-fluorophenylboronic acid, boronic acid,b-3-ethoxy-5-fluorophenyl, 3-ethoxy-5-fluorophenylboronicacid, acmc-209q3b, 3-ethoxy-5-fluoro-benzeneboronic acid, 3-ethoxy-5-fluoro-phenyl boronic acid, 3-ethoxy-5-fluorobenzeneboronicacid, 3-ethoxy-5-fluoranyl-phenyl boronic acid | |
(3-ethoxy-5-fluorophenyl)boronic acid |
Specifications
850589-53-6 | |
White | |
MFCD07363779 | |
3-ethoxy-5-fluorobenzeneboronic acid, 3-ethoxy-5-fluorophenyl boronic acid, 5-ethoxy-3-fluorophenylboronic acid, boronic acid,b-3-ethoxy-5-fluorophenyl, 3-ethoxy-5-fluorophenylboronicacid, acmc-209q3b, 3-ethoxy-5-fluoro-benzeneboronic acid, 3-ethoxy-5-fluoro-phenyl boronic acid, 3-ethoxy-5-fluorobenzeneboronicacid, 3-ethoxy-5-fluoranyl-phenyl boronic acid | |
CCOC1=CC(F)=CC(=C1)B(O)O | |
183.97 | |
183.97 | |
3-Ethoxy-5-fluorophenylboronic Acid (contains varying amounts of Anhydride) |
158°C | |
C8H10BFO3 | |
200 mg | |
OVVBLFYHZAMJIK-UHFFFAOYSA-N | |
(3-ethoxy-5-fluorophenyl)boronic acid | |
44558175 | |
Crystalline Powder |
Safety and Handling
TSCA : No