missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethoxy-4-methoxybenzonitrile, TCI America™

Manufacturer:  TCI America E110925G

 View more versions of this product

Catalog No. E110925G



Specifications

Specifications

3-Ethoxy-4-methoxybenzonitrile
72°C
60758-86-3
MFCD02256160
XTIINWPNAMHVDG-UHFFFAOYSA-N
3-ethoxy-4-methoxybenzonitrile
3934769
Crystal-Powder at 20°C
White-Yellow
25g
C10H11NO2
3439
CCOC1=C(C=CC(=C1)C#N)OC
177.203
177.20
≥98.0% (GC)

Chemical Identifiers

60758-86-3
177.203
XTIINWPNAMHVDG-UHFFFAOYSA-N
3-ethoxy-4-methoxybenzonitrile
C10H11NO2
MFCD02256160
3934769
CCOC1=C(C=CC(=C1)C#N)OC
Safety and Handling

Safety and Handling

TSCA : No

Documents