missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Ethoxy-4-methoxybenzaldehyde, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2476214
Description
3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Air sensitive. Store in a tightly sealed container. Store away from oxidizing agents, air.
Specifications
3-Ethoxy-4-methoxybenzaldehyde | |
51°C to 53°C | |
MFCD00010128 | |
Air Sensitive | |
Soluble in methanol. | |
CCOC1=C(C=CC(=C1)C=O)OC | |
180.203 | |
180.2 |
1131-52-8 | |
C10H12O3 | |
25 g | |
3-ethoxy-4-methoxy-benzaldehyde, benzaldehyde, 3-ethoxy-4-methoxy, pubchem13135, 3-ethoxy-p-anisaldehyde, acmc-1ca82, 3-ethoxy-4methoxybenzaldehyde, 3ethoxy-4-methoxybenzaldehyde, ksc183c6j, 3-ethoxy-4-methoxybenzaldehyd, 4-methoxy-3-ethoxybenzaldehyde | |
VAMZHXWLGRQSJS-UHFFFAOYSA-N | |
3-ethoxy-4-methoxybenzaldehyde | |
244728 | |
99% |
Chemical Identifiers
1131-52-8 | |
180.203 | |
VAMZHXWLGRQSJS-UHFFFAOYSA-N | |
244728 | |
CCOC1=C(C=CC(=C1)C=O)OC |
C10H12O3 | |
MFCD00010128 | |
3-ethoxy-4-methoxy-benzaldehyde, benzaldehyde, 3-ethoxy-4-methoxy, pubchem13135, 3-ethoxy-p-anisaldehyde, acmc-1ca82, 3-ethoxy-4methoxybenzaldehyde, 3ethoxy-4-methoxybenzaldehyde, ksc183c6j, 3-ethoxy-4-methoxybenzaldehyd, 4-methoxy-3-ethoxybenzaldehyde | |
3-ethoxy-4-methoxybenzaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only