missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Ethoxy-4-hydroxybenzaldehyde, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 156030050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Ethoxy-4-hydroxybenzaldehyde | |
121-32-4 | |
100.0 | |
96% min. (GC) | |
C9H10O3 | |
MFCD00006944 | |
08,256 | |
ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal | |
CBOQJANXLMLOSS-UHFFFAOYSA-N | |
3-ethoxy-4-hydroxybenzaldehyde | |
8467 | |
166.18 |
97% | |
96.0 | |
Authentic | |
Glass bottle | |
C2H5OC6H3(OH)CHO | |
5g | |
15,3908 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,glycerol and prpylene,glycol | |
CCOC1=CC(C=O)=CC=C1O | |
166.18 | |
CHEBI:48408 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 204-464-7
RTECSNumber : CU6125000
TSCA : TSCA
RUO â Research Use Only