Learn More
3-Ethoxy-4-hydroxybenzaldehyde 97%, ACROS Organics™
Manufacturer: Acros Organics AC156035000
Specifications
3-Ethoxy-4-hydroxybenzaldehyde | |
Glass bottle | |
76.0°C to 79.0°C | |
97% | |
96.0 | |
97% | |
C2H5OC6H3(OH)CHO | |
ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal | |
CCOC1=C(C=CC(=C1)C=O)O | |
166.176 | |
CHEBI:48408 | |
Fine Crystalline Powder |
Authentic | |
White | |
500g | |
121-32-4 | |
100.0 | |
C9H10O3 | |
MFCD00006944 | |
CBOQJANXLMLOSS-UHFFFAOYSA-N | |
3-ethoxy-4-hydroxybenzaldehyde | |
8467 | |
166.18 | |
97% |
Chemical Identifiers
121-32-4 | |
166.176 | |
CBOQJANXLMLOSS-UHFFFAOYSA-N | |
8467 | |
3-ethoxy-4-hydroxybenzaldehyde |
C9H10O3 | |
MFCD00006944 | |
ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal | |
CHEBI:48408 | |
CCOC1=C(C=CC(=C1)C=O)O |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
LIGHT SENSITIVE
EINECSNumber : 204-464-7
RTECSNumber : CU6125000
TSCA : TSCA