Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxy-2-nitropyridine, 98%

$223.00 - $845.30

Chemical Identifiers

Chemical Name or Material 3-Ethoxy-2-nitropyridine
CAS 74037-50-6
Molecular Formula C7H8N2O3
Formula Weight 168.15
MDL Number MFCD00006257
Beilstein 141592
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1437106

 
alfa aesar™
A1437106
5g Each for $223.00

AAA1437109

 
alfa aesar™
A1437109
10g Each for $409.84

AAA1437114

 
alfa aesar™
A1437114
25g Each for $845.30
Description & Specifications

Specifications

Formula Weight 168.15
MDL Number MFCD00006257
CAS 74037-50-6
Melting Point 28° to 32°C
RTECS Number UT3001500
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H8N2O3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 156° to 158°C (15mmHg)
Quantity 5g
Flash Point >100°C (212°F)
Beilstein 141592
Assay 98%
Chemical Name or Material 3-Ethoxy-2-nitropyridine
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.