missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Ethoxy-2-cyclohexen-1-one, TCI America™

Manufacturer:  TCI America E038425ML

 View more versions of this product

Catalog No. E038425ML



Specifications

Specifications

3-Ethoxy-2-cyclohexen-1-one
Yellow
5323-87-5
MFCD00001580
JWCFJPLIRVYENQ-UHFFFAOYSA-N
3-ethoxycyclohex-2-en-1-one
79216
Clear Liquid at 20°C
119°C
25mL
C8H12O2
3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone
CCOC1=CC(=O)CCC1
140.182
140.18
≥98.0% (GC)

Chemical Identifiers

5323-87-5
140.182
JWCFJPLIRVYENQ-UHFFFAOYSA-N
79216
CCOC1=CC(=O)CCC1
C8H12O2
MFCD00001580
3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone
3-ethoxycyclohex-2-en-1-one
Safety and Handling

Safety and Handling

TSCA : No

Documents