Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxy-2,6-difluorobenzoic acid, 97%

$114.22 - $114.22

Chemical Identifiers

Chemical Name or Material 3-Ethoxy-2,6-difluorobenzoic acid
Molecular Formula C9H8F2O3
Formula Weight 202.15
MDL Number MFCD11519368
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3193603

 
alfa aesar™
H3193603
1g Each for $114.22

AAH3193606

 
alfa aesar™
H3193606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.15
Chemical Name or Material 3-Ethoxy-2,6-difluorobenzoic acid
Assay 97%
Molecular Formula C9H8F2O3
MDL Number MFCD11519368
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 162° to 164°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.