Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxy-2,6-difluoroaniline, 97%

Chemical Identifiers

Chemical Name or Material 3-Ethoxy-2,6-difluoroaniline
Molecular Formula C8H9F2NO
Formula Weight 173.16
MDL Number MFCD11519309
UN Number UN2810
Synonym 2,6-Difluoro-m-phenetidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3273203

 
alfa aesar™
H3273203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3273206

 
alfa aesar™
H3273206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.16
Chemical Name or Material 3-Ethoxy-2,6-difluoroaniline
Assay 97%
Molecular Formula C8H9F2NO
MDL Number MFCD11519309
UN Number UN2810
Synonym 2,6-Difluoro-m-phenetidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.