Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Ethoxy-2,4-difluorobenzaldehyde, 97%

$68.50 - $275.53

Chemical Identifiers

Chemical Name or Material 3-Ethoxy-2,4-difluorobenzaldehyde
Molecular Formula C9H8F2O2
Formula Weight 186.16
MDL Number MFCD09258722
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3263103

 
alfa aesar™
H3263103
1g Each for $68.50

AAH3263106

 
alfa aesar™
H3263106
5g Each for $275.53
Description & Specifications

Specifications

Formula Weight 186.16
Chemical Name or Material 3-Ethoxy-2,4-difluorobenzaldehyde
Assay 97%
Molecular Formula C9H8F2O2
MDL Number MFCD09258722
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.