Please login to your online account to display your discounted pricing

3-endo-Aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-endo-Aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid
Name Note 98%
CAS 88330-29-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8 H11 N O2
Formula Weight 153.18
MDL Number MFCD00143119
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC265670010

  SDS
thermo fisher scientific
265670010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 153.18
Quantity 1g
Color Beige
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
CAS 88330-29-4
Chemical Name or Material 3-endo-Aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylic acid
Packaging Glass bottle
Name Note 98%
Assay Percent Range 97%
Molecular Formula C8 H11 N O2
MDL Number MFCD00143119
Infrared Spectrum Authentic