Please login to your online account to display your discounted pricing

3-Dodecylthiophene, 97%, Acros Organics


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  333820010

 View more versions of this product

Catalog No. AC333820010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 252.56
Physical Form Liquid
Quantity 1g
Color Clear colorless to yellow
TSCA TSCA
Density 0.9020g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-Dodecylthiophene
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C16H28S
Packaging Glass bottle
MDL Number MFCD00130121
CAS 104934-52-3
Refractive Index 1.4870 to 1.4890 (20°C, 589nm)

Specifications

Chemical Name or Material 3-Dodecylthiophene
CAS 104934-52-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C16H28S
Formula Weight 252.56
MDL Number MFCD00130121