missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Dodecylthiophene, 97%, Acros Organics

Chemical Identifiers

CAS 104934-52-3
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C16H28S
Formula Weight 252.56
MDL Number MFCD00130121
Chemical Name or Material 3-Dodecylthiophene
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC333820010 SDS Thermo Fisher Scientific
333820010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

104934-52-3
100.0
C16H28S
MFCD00130121
≥96.0%
96.0
97%
252.56
3-Dodecylthiophene

Specifications

252.56
1g
0.9020g/mL
100.0
Glass bottle
104934-52-3
C16H28S
3-Dodecylthiophene
Liquid
Clear colorless to yellow
96.0
Authentic
1.4870 to 1.4890 (20°C, 589nm)
97%
MFCD00130121
≥96.0%

Safety and Handling

TSCA : TSCA