missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Dimethylcarbamoyl)-2-fluorobenzeneboronic acid, 95%

Manufacturer:  Alfa Aesar™ H5608703

 View more versions of this product

Catalog No. AAH5608703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Notes
Reacts with strong oxidizing agents.
Specifications

Specifications

3-(Dimethylcarbamoyl)-2-fluorobenzeneboronic acid
1g
C9H11BFNO3
ICJWWRWLFPUVJI-UHFFFAOYSA-N
[3-(dimethylcarbamoyl)-2-fluorophenyl]boronic acid
57497310
Solid
White
1451391-88-0
2-fluoro-3-n,n-dimethylaminocarbonyl phenylboronic acid
B(C1=C(C(=CC=C1)C(=O)N(C)C)F)(O)O
210.999
210.99
95%

Chemical Identifiers

1451391-88-0
210.999
2-fluoro-3-n,n-dimethylaminocarbonyl phenylboronic acid
[3-(dimethylcarbamoyl)-2-fluorophenyl]boronic acid
C9H11BFNO3
ICJWWRWLFPUVJI-UHFFFAOYSA-N
57497310
B(C1=C(C(=CC=C1)C(=O)N(C)C)F)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents