Learn More
3-Dimethylaminopropylamine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 115900025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Dimethylaminopropylamine | |
98.5 | |
0.8100g/mL | |
32°C | |
98.5% min. (GC) | |
C5H14N2 | |
(CH3)2N(CH2)3NH2 | |
2.5L | |
01,274 | |
3-dimethylaminopropylamine, 3-dimethylamino propylamine, 3-dimethylamino-1-propylamine, n,n-dimethyl-1,3-propanediamine, 1,3-propanediamine, n,n-dimethyl, n,n-dimethyltrimethylenediamine, 3-aminopropyldimethylamine, n,n-dimethylpropane-1,3-diamine, 3-aminopropyl dimethylamine, dimethylaminopropylamine | |
IUNMPGNGSSIWFP-UHFFFAOYSA-N | |
N',N'-dimethylpropane-1,3-diamine | |
7993 | |
102.18 |
109-55-7 | |
100.0 | |
133.0°C | |
Authentic | |
Glass bottle | |
1.4340 to 1.4360 | |
MFCD00008216 | |
04, III, 554 | |
0.81 | |
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents | |
CN(C)CCCN | |
102.18 | |
1.6 mPa.s (20°C) | |
99% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Harmful in contact with skin.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 203-680-9
RUO â Research Use Only