Learn More
3-Dimethylaminopropyl chloride, 97%, stab. with 0.5% oxalic acid, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6537106
Description
It is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in chlorpromazine, anti-inflammatory pain static, propylene clomipramine work, doxpin and amy for butyl intermediates.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
3-Dimethylaminopropyl chloride | |
168°C | |
0.929 g/mL | |
1.432 | |
MFCD00044496 | |
605294 | |
3-chloro-1-n,n-dimethyl propylamine, dimethylaminopropyl chloride, 3-chloropropyl dimethylamine, 3-chloropropyl dimethyl amine, 1-dimethylamino-3-chloropropane, 1-propanamine, 3-chloro-n,n-dimethyl, n-3-chloropropyl dimethylamine, 3-chloro-propyl-dimethyl-amine, 3-dimethylaminopropyl chloride, 3-dimethylaminopropylchloride | |
NYYRRBOMNHUCLB-UHFFFAOYSA-N | |
3-chloro-N,N-dimethylpropan-1-amine | |
66960 | |
97% |
109-54-6 | |
Colorless | |
C5H12ClN | |
Cl(CH2)3N(CH3)2 | |
5 g | |
Air sensitive | |
Slightly soluble in water. | |
CN(C)CCCCl | |
121.608 | |
121.61 | |
Liquid |
Chemical Identifiers
109-54-6 | |
121.608 | |
NYYRRBOMNHUCLB-UHFFFAOYSA-N | |
66960 | |
CN(C)CCCCl |
C5H12ClN | |
MFCD00044496 | |
3-chloro-1-n,n-dimethyl propylamine, dimethylaminopropyl chloride, 3-chloropropyl dimethylamine, 3-chloropropyl dimethyl amine, 1-dimethylamino-3-chloropropane, 1-propanamine, 3-chloro-n,n-dimethyl, n-3-chloropropyl dimethylamine, 3-chloro-propyl-dimethyl-amine, 3-dimethylaminopropyl chloride, 3-dimethylaminopropylchloride | |
3-chloro-N,N-dimethylpropan-1-amine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 203-679-3
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only