missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Dimethylaminopropiophenone hydrochloride, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 115890250
Specifications
3-Dimethylaminopropiophenone hydrochloride | |
152.0°C to 155.0°C | |
99% | |
98.5 | |
99% | |
C6H5COCH2CH2N(CH3)2·HCl | |
14, II, 38 | |
DKNDBIIKSJWQFL-UHFFFAOYSA-N | |
3-(dimethylamino)-1-phenylpropan-1-one;hydrochloride | |
92844 | |
99% |
Glass bottle | |
25g | |
879-72-1 | |
100.0 | |
C11H15NO·HCl | |
MFCD00012481 | |
1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride 1:1, 3-dimethylamino propiophenone hydrochloride, 3-dimethylamino-1-phenylpropan-1-one hydrochloride, 3-dimethylaminopropiophenone hydrochloride, beta-dimethylaminopropiophenone hydrochloride, dimethylaminoethylphenylketone hydrochloride, nsc 89, propiophenone, 3-dimethylamino-, hydrochloride, wln: 1n1&2vr &gh | |
CN(C)CCC(=O)C1=CC=CC=C1.Cl | |
213.71 | |
213.71 |
Chemical Identifiers
879-72-1 | |
213.71 | |
DKNDBIIKSJWQFL-UHFFFAOYSA-N | |
92844 | |
CN(C)CCC(=O)C1=CC=CC=C1.Cl |
C11H15NO·HCl | |
MFCD00012481 | |
1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride 1:1, 3-dimethylamino propiophenone hydrochloride, 3-dimethylamino-1-phenylpropan-1-one hydrochloride, 3-dimethylaminopropiophenone hydrochloride, beta-dimethylaminopropiophenone hydrochloride, dimethylaminoethylphenylketone hydrochloride, nsc 89, propiophenone, 3-dimethylamino-, hydrochloride, wln: 1n1&2vr &gh | |
3-(dimethylamino)-1-phenylpropan-1-one;hydrochloride |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 212-909-1
RTECSNumber : UH0600000
TSCA : TSCA