missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Dimethylaminopropiophenone hydrochloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115890050

 View more versions of this product

Catalog No. AC115890050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Dimethylaminopropiophenone hydrochloride
152.0°C to 155.0°C
99%
98.5
99%
C6H5COCH2CH2N(CH3)2·HCl
14, II, 38
DKNDBIIKSJWQFL-UHFFFAOYSA-N
3-(dimethylamino)-1-phenylpropan-1-one;hydrochloride
92844
99%
Glass bottle
5g
879-72-1
100.0
C11H15NO·HCl
MFCD00012481
1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride 1:1, 3-dimethylamino propiophenone hydrochloride, 3-dimethylamino-1-phenylpropan-1-one hydrochloride, 3-dimethylaminopropiophenone hydrochloride, beta-dimethylaminopropiophenone hydrochloride, dimethylaminoethylphenylketone hydrochloride, nsc 89, propiophenone, 3-dimethylamino-, hydrochloride, wln: 1n1&2vr &gh
CN(C)CCC(=O)C1=CC=CC=C1.Cl
213.71
213.71

Chemical Identifiers

879-72-1
213.71
DKNDBIIKSJWQFL-UHFFFAOYSA-N
92844
CN(C)CCC(=O)C1=CC=CC=C1.Cl
C11H15NO·HCl
MFCD00012481
1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride, 1-propanone, 3-dimethylamino-1-phenyl-, hydrochloride 1:1, 3-dimethylamino propiophenone hydrochloride, 3-dimethylamino-1-phenylpropan-1-one hydrochloride, 3-dimethylaminopropiophenone hydrochloride, beta-dimethylaminopropiophenone hydrochloride, dimethylaminoethylphenylketone hydrochloride, nsc 89, propiophenone, 3-dimethylamino-, hydrochloride, wln: 1n1&2vr &gh
3-(dimethylamino)-1-phenylpropan-1-one;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 212-909-1

RTECSNumber : UH0600000

TSCA : TSCA

SDS Documents