missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Dimethylaminomethyl)-5-nitroindole, 95%

$106.55 - $1,153.11

Chemical Identifiers

CAS 3414-64-0
Molecular Formula C11H13N3O2
Molecular Weight (g/mol) 219.244
MDL Number MFCD00544197
InChI Key SVNVKGIWGOHXPW-UHFFFAOYSA-N
Synonym 1h-indole-3-methanamine, n,n-dimethyl-5-nitro, 1h-indole-3-methanamine,n-dimethyl-5-nitro, 3-dimethylamino methyl-5-nitroindole, 3-dimethylaminomethyl-5-nitroindole, 4-22-00-04317 beilstein handbook reference, 5-nitrogramine, dimethyl 5-nitro-1h-indol-3-yl methyl amine, indole, 3-dimethylamino methyl-5-nitro, n,n-dimethyl-1-5-nitro-1h-indol-3-yl methanamine
PubChem CID 18882
IUPAC Name N,N-dimethyl-1-(5-nitro-1H-indol-3-yl)methanamine
SMILES CN(C)CC1=CNC2=C1C=C(C=C2)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63939MD
View Documents
Alfa Aesar™
H63939MD
250mg
Each for $106.55
 
AAH6393903
View Documents
Alfa Aesar™
H6393903
1g
Each for $252.05
 
AAH6393906
View Documents
Alfa Aesar™
H6393906
5g
Each for $1,153.11
 
Description

Description

Specifications

Chemical Identifiers

3414-64-0
219.244
SVNVKGIWGOHXPW-UHFFFAOYSA-N
18882
CN(C)CC1=CNC2=C1C=C(C=C2)[N+](=O)[O-]
C11H13N3O2
MFCD00544197
1h-indole-3-methanamine, n,n-dimethyl-5-nitro, 1h-indole-3-methanamine,n-dimethyl-5-nitro, 3-dimethylamino methyl-5-nitroindole, 3-dimethylaminomethyl-5-nitroindole, 4-22-00-04317 beilstein handbook reference, 5-nitrogramine, dimethyl 5-nitro-1h-indol-3-yl methyl amine, indole, 3-dimethylamino methyl-5-nitro, n,n-dimethyl-1-5-nitro-1h-indol-3-yl methanamine
N,N-dimethyl-1-(5-nitro-1H-indol-3-yl)methanamine

Specifications

250mg
C11H13N3O2
216692
SVNVKGIWGOHXPW-UHFFFAOYSA-N
N,N-dimethyl-1-(5-nitro-1H-indol-3-yl)methanamine
18882
Solid
3-(Dimethylaminomethyl)-5-nitroindole
3414-64-0
MFCD00544197
1h-indole-3-methanamine, n,n-dimethyl-5-nitro, 1h-indole-3-methanamine,n-dimethyl-5-nitro, 3-dimethylamino methyl-5-nitroindole, 3-dimethylaminomethyl-5-nitroindole, 4-22-00-04317 beilstein handbook reference, 5-nitrogramine, dimethyl 5-nitro-1h-indol-3-yl methyl amine, indole, 3-dimethylamino methyl-5-nitro, n,n-dimethyl-1-5-nitro-1h-indol-3-yl methanamine
CN(C)CC1=CNC2=C1C=C(C=C2)[N+](=O)[O-]
219.244
219.24
95%
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : NL7710000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Product Certifications