missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Dimethylaminomethyl)-5-nitroindole, 95%

Manufacturer:  Alfa Aesar™ H63939MD

 View more versions of this product

Catalog No. AAH63939MD



Specifications

Specifications

3-(Dimethylaminomethyl)-5-nitroindole
3414-64-0
MFCD00544197
5-nitrogramine, n,n-dimethyl-1-5-nitro-1h-indol-3-yl methanamine, 1h-indole-3-methanamine, n,n-dimethyl-5-nitro, indole, 3-dimethylamino methyl-5-nitro, 3-dimethylamino methyl-5-nitroindole, indole, 3-dimethylamino methyl-5-nitro, dimethyl 5-nitro-1h-indol-3-yl methyl amine, 4-22-00-04317 beilstein handbook reference, 3-dimethylaminomethyl-5-nitroindole, 1h-indole-3-methanamine,n-dimethyl-5-nitro
CN(C)CC1=CNC2=C1C=C(C=C2)[N+](=O)[O-]
219.244
219.24
95%
250mg
C11H13N3O2
216692
SVNVKGIWGOHXPW-UHFFFAOYSA-N
N,N-dimethyl-1-(5-nitro-1H-indol-3-yl)methanamine
18882
Solid

Chemical Identifiers

3414-64-0
219.244
SVNVKGIWGOHXPW-UHFFFAOYSA-N
18882
CN(C)CC1=CNC2=C1C=C(C=C2)[N+](=O)[O-]
C11H13N3O2
MFCD00544197
5-nitrogramine, n,n-dimethyl-1-5-nitro-1h-indol-3-yl methanamine, 1h-indole-3-methanamine, n,n-dimethyl-5-nitro, indole, 3-dimethylamino methyl-5-nitro, 3-dimethylamino methyl-5-nitroindole, indole, 3-dimethylamino methyl-5-nitro, dimethyl 5-nitro-1h-indol-3-yl methyl amine, 4-22-00-04317 beilstein handbook reference, 3-dimethylaminomethyl-5-nitroindole, 1h-indole-3-methanamine,n-dimethyl-5-nitro
N,N-dimethyl-1-(5-nitro-1H-indol-3-yl)methanamine
Safety and Handling

Safety and Handling

RTECSNumber : NL7710000

storageNote1 : 2° to 8°C

Documents