Learn More
3-Dimethylaminobenzoic acid, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 115751000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Dimethylaminobenzoic acid | |
99-64-9 | |
100.0 | |
99% | |
C9H11NO2 | |
MFCD00002497 | |
14, 392 | |
Solubility in water: insoluble. | |
CN(C)C1=CC=CC(=C1)C(=O)O | |
165.19 | |
165.19 |
99% | |
98.5 | |
Authentic | |
Plastic bottle | |
(CH3)2NC6H4CO2H | |
100g | |
3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid | |
NEGFNJRAUMCZMY-UHFFFAOYSA-N | |
3-(dimethylamino)benzoic acid | |
66837 | |
99% |
Chemical Identifiers
99-64-9 | |
165.19 | |
NEGFNJRAUMCZMY-UHFFFAOYSA-N | |
66837 | |
CN(C)C1=CC=CC(=C1)C(=O)O |
C9H11NO2 | |
MFCD00002497 | |
3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid | |
3-(dimethylamino)benzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 202-775-2
TSCA : TSCA
RUO â Research Use Only