Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Dimethylaminobenzoic acid, 98%

$57.11 - $232.04

Chemical Identifiers

Chemical Name or Material 3-Dimethylaminobenzoic acid
CAS 99-64-9
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD00002497
Beilstein 2208586
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1361222

  SDS
alfa aesar™
A1361222
100g Each for $57.11

AAA1361236

  SDS
alfa aesar™
A1361236
500g Each for $232.04
Description & Specifications

Specifications

Formula Weight 165.19
Chemical Name or Material 3-Dimethylaminobenzoic acid
TSCA TSCA
Assay 98%
Molecular Formula C9H11NO2
Melting Point 148° to 153°C
MDL Number MFCD00002497
CAS 99-64-9
Beilstein 2208586
Quantity 100g
Odor Odorless

Applications
3-Dimethylaminobenzoic acid is used as a benzoic acid derivative.

Solubility
Soluble in methanol (50 mg/ml). Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Incompatible with oxidizing agents.