Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(Dimethylamino)propionitrile, 98%, Electrophoresis Grade

Chemical Identifiers

Chemical Name or Material 3-(Dimethylamino)propionitrile
CAS 1738-25-6
Purity Grade Electrophoresis Grade
Linear Formula (CH3)2NCH2CH2CN
Formula Weight 98.15
MDL Number MFCD00001957
Beilstein 773779
Synonym DMAPN
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66058AK

  SDS
alfa aesar™
J66058AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 60° to 61°C
Formula Weight 98.15
Chemical Name or Material 3-(Dimethylamino)propionitrile
Physical Form Liquid
MDL Number MFCD00001957
Synonym DMAPN
TSCA TSCA
Quantity 250mL
Purity Grade Electrophoresis Grade
Assay 98%
Linear Formula (CH3)2NCH2CH2CN
CAS 1738-25-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H312-H315-H319-H335-H227
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UG1575000
Density 0.870g/mL
Melting Point −44°C
Color Colorless to yellow
Beilstein 773779
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H312-H315-H319-H335-H227
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.