Please login to your online account to display your discounted pricing

3-(Dimethylamino)propionitrile, 98%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  215451000

 View more versions of this product

Catalog No. AC215451000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 98.15
Quantity 100mL
Color Clear colorless to yellow
Beilstein 04, III, 1265
EINECS Number 217-090-4
TSCA TSCA
RTECS Number UG1575000
Density 0.87g/mL
CAS Max % 100.0
Chemical Name or Material 3-(Dimethylamino)propionitrile
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C5H10N2
Linear Formula (CH3)2NCH2CH2CN
MDL Number MFCD00001957
CAS 1738-25-6
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-(Dimethylamino)propionitrile
Name Note 98%
CAS 1738-25-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H10N2
Linear Formula (CH3)2NCH2CH2CN
Formula Weight 98.15
MDL Number MFCD00001957
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents