missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(Dimethylamino)phenylboronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444950010
Specifications
3-(Dimethylamino)phenylboronic acid pinacol ester | |
53°C to 55°C | |
325142-87-8 | |
C14H22BNO2 | |
BIJRPONWUBCGES-UHFFFAOYSA-N | |
N,N-dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
17998909 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
3-dimethylamino phenylboronic acid pinacol ester, 3-n,n-dimethylamino phenylboronic acid pinacol ester, 3-n,n-dimethylamino phenylboronic acid, pinacol ester, benzenamine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, dimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline, n,n-dimethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N(C)C | |
247.14 | |
247.14 |
Chemical Identifiers
325142-87-8 | |
247.14 | |
3-dimethylamino phenylboronic acid pinacol ester, 3-n,n-dimethylamino phenylboronic acid pinacol ester, 3-n,n-dimethylamino phenylboronic acid, pinacol ester, benzenamine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, dimethyl 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl amine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzenamine, n,n-dimethyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline, n,n-dimethyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline | |
N,N-dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
C14H22BNO2 | |
BIJRPONWUBCGES-UHFFFAOYSA-N | |
17998909 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N(C)C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning