Please login to your online account to display your discounted pricing

3-(Dimethylamino)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Dimethylamino)phenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C14H22BNO2
Formula Weight 247.14
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444950050

 
thermo fisher scientific
AC444950050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444950010

 
thermo fisher scientific
AC444950010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.14
Quantity 5g
Chemical Name or Material 3-(Dimethylamino)phenylboronic acid pinacol ester
Assay Percent Range 97%
CAS 325142-87-8
Molecular Formula C14H22BNO2