missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Dimethylamino-2-methylpropyl chloride hydrochloride, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC166795000

 View more versions of this product

Catalog No. AC166795000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Dimethylamino-2-methylpropyl chloride hydrochloride
Glass bottle
169°C to 173°C
4261-67-0
100.0
C6H15Cl2N
MFCD00012514
3-dimethylamino-2-methylpropyl chloride hydrochloride, 3-chloro-n,n,2-trimethylpropan-1-amine hydrochloride, 3-chloro-2-methylpropyl dimethyl ammonium chloride, 3-chloro-2-methylpropyl dimethylamine hydrochloride, acmc-20an51, 3-dimethylamino-2-methylpropyl chloride hcl, 3-chloro-n,n,2-trimethylpropylamine hydrochloride, 2-methyl-3-dimethylaminopropylchloride hydrochloride, 3-dimethylamino-2-methylpropylchloride hydrochloride, 1-dimethylamino-2-methyl-3-chloropropane hydrochloride
CC(CN(C)C)CCl.Cl
172.093
172.1
≥97.5%
Authentic
White
500g
97.5
98%
(CH3)2NCH2CH(CH3)CH2Cl·HCl
04, IV, 655
SOMIBONUMGNAEP-UHFFFAOYSA-N
3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride
3083841
Powder

Chemical Identifiers

4261-67-0
172.093
SOMIBONUMGNAEP-UHFFFAOYSA-N
3083841
CC(CN(C)C)CCl.Cl
C6H15Cl2N
MFCD00012514
3-dimethylamino-2-methylpropyl chloride hydrochloride, 3-chloro-n,n,2-trimethylpropan-1-amine hydrochloride, 3-chloro-2-methylpropyl dimethyl ammonium chloride, 3-chloro-2-methylpropyl dimethylamine hydrochloride, acmc-20an51, 3-dimethylamino-2-methylpropyl chloride hcl, 3-chloro-n,n,2-trimethylpropylamine hydrochloride, 2-methyl-3-dimethylaminopropylchloride hydrochloride, 3-dimethylamino-2-methylpropylchloride hydrochloride, 1-dimethylamino-2-methyl-3-chloropropane hydrochloride
3-chloro-N,N,2-trimethylpropan-1-amine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents